siRNA Details
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106468.1 | NP_001099938.1 | 15 | 3311 | ||
XM_006233262.1 | XP_006233324.1 | 16 | 3435 | |||
GenBank mRNA | FQ222386.1 | - | 16 | 3440 |
