siRNA Details
Gene Symbol: Ints12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007640.1 | NP_001007641.1 | 2 | 275 | ||
XM_006233320.2 | XP_006233382.1 | 3 | 1405 | |||
XM_006233321.2 | XP_006233383.1 | 3 | 391 | |||
GenBank mRNA | BC079404.1 | - | 2 | 275 | 7112817 | |
FQ212179.1 | - | 3 | 366 |
