siRNA Details
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011953.1 | NP_001011953.1 | 6, 7 | 601 | ||
XM_008761549.1 | XP_008759771.1 | 6, 7 | 627 | |||
GenBank mRNA | BC083636.1 | - | 6, 7 | 601 | 7134753 | |
FQ219584.1 | - | 1, 2 | 78 |
