siRNA Details
Gene Symbol: Prpf40a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106480.1 | NP_001099950.1 | 23 | 2498 | ||
XM_006234164.2 | XP_006234226.1 | 23 | 2823 | |||
XM_008761709.1 | XP_008759931.1 | 23 | 2962 | |||
XM_008761710.1 | XP_008759932.1 | 22 | 2909 | |||
XM_008761711.1 | XP_008759933.1 | 23 | 2835 | |||
XM_008761712.1 | XP_008759934.1 | 22 | 2782 | |||
XM_008761713.1 | XP_008759935.1 | 22 | 2286 |
