siRNA Details
Gene Symbol: Prpf40a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106480.1 | NP_001099950.1 | 13 | 1299 | ||
XM_006234164.2 | XP_006234226.1 | 13 | 1624 | |||
XM_008761709.1 | XP_008759931.1 | 13 | 1751 | |||
XM_008761710.1 | XP_008759932.1 | 12 | 1698 | |||
XM_008761711.1 | XP_008759933.1 | 13 | 1624 | |||
XM_008761712.1 | XP_008759934.1 | 12 | 1571 | |||
XM_008761713.1 | XP_008759935.1 | 12 | 1075 |
