siRNA Details
Gene Symbol: Sestd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134514.2 | NP_001127986.2 | 17 | 1973 | ||
XM_006234402.2 | XP_006234464.1 | 18 | 2253 | |||
XM_006234404.2 | XP_006234466.1 | 17 | 2163 | |||
XM_008761892.1 | XP_008760114.1 | 18 | 2179 | |||
XM_008761893.1 | XP_008760115.1 | 17 | 2089 | |||
GenBank mRNA | BC169110.1 | - | 17 | 1973 | 7445081 |
