siRNA Details
Gene Symbol: Serping1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199093.1 | NP_954524.1 | 6 | 1202 | ||
XM_006234447.2 | XP_006234509.1 | 7 | 1277 | |||
GenBank mRNA | BC061860.1 | - | 6 | 1202 | 5598525 | |
FQ219227.1 | - | 5 | 907 |
