siRNA Details
Gene Symbol: Slc37a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031589.2 | NP_113777.2 | 4 | 744 | ||
XM_008766151.1 | XP_008764373.1 | 3 | 505 | |||
XM_008766152.1 | XP_008764374.1 | 3 | 570 | |||
XM_008766153.1 | XP_008764375.1 | 3 | 505 | |||
GenBank mRNA | AF080468.1 | - | 3 | 574 | ||
BC070889.1 | - | 4 | 744 | 7098680 |
