siRNA Details
Gene Symbol: Fdft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019238.2 | NP_062111.1 | 7 | 991 | ||
XM_008770746.1 | XP_008768968.1 | 6 | 3638 | |||
GenBank mRNA | BC081810.1 | - | 7 | 991 | 7188993 | |
FQ213323.1 | - | 7 | 979 | |||
FQ219372.1 | - | 7 | 959 | |||
M95591.1 | - | 7 | 1009 |
