siRNA Details
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106490.1 | NP_001099960.1 | 21 | 2316 | ||
XM_006234503.2 | XP_006234565.1 | 23 | 2372 | |||
XM_006234504.1 | XP_006234566.1 | 23 | 2372 |
