siRNA Details
Gene Symbol: Commd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033692.1 | NP_001028864.1 | 5 | 390 | ||
XM_006234637.2 | XP_006234699.1 | 5 | 425 | |||
GenBank mRNA | BC101873.1 | - | 5 | 390 | 5621281 | |
EU000469.1 | - | 5 | 386 |
