siRNA Details
Gene Symbol: Ehf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106493.2 | NP_001099963.1 | 5 | 552 | ||
XM_006234639.2 | XP_006234701.1 | 5 | 3051 | |||
XM_006234641.2 | XP_006234703.1 | 5 | 626 | |||
GenBank mRNA | BC166742.1 | - | 5 | 552 | 7937842 |
