siRNA Details
Gene Symbol: Dtwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013921.1 | NP_001013943.1 | 3 | 762 | ||
XM_006234910.2 | XP_006234972.1 | 4 | 741 | |||
XM_006234914.2 | XP_006234976.1 | 4 | 540 | |||
XM_006234915.2 | XP_006234977.1 | 4 | 675 | |||
XM_008762154.1 | XP_008760376.1 | 4 | 1553 | |||
GenBank mRNA | BC079068.1 | - | 3 | 762 | 7121624 |
