siRNA Details
Gene Symbol: Gpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168529.1 | NP_001162001.1 | 11 | 1275 | ||
XM_006234956.2 | XP_006235018.1 | 14 | 1673 | |||
XM_008762158.1 | XP_008760380.1 | 12 | 1936 |
