siRNA Details
Gene Symbol: Nphp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106506.1 | NP_001099976.1 | 14, 15 | 2193 | ||
XM_006234961.2 | XP_006235023.1 | 15, 16 | 2437 | |||
XM_006234963.2 | XP_006235025.1 | 15, 16 | 2434 | |||
XM_006234964.2 | XP_006235026.1 | 15, 16 | 2434 | |||
XM_006234965.2 | XP_006235027.1 | 15, 16 | 2431 | |||
GenBank mRNA | BC168839.1 | - | 10, 11 | 1157 |
