siRNA Details
Gene Symbol: Ndufaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126371.1 | NP_001119843.1 | 2 | 271 | ||
XM_006235097.2 | XP_006235159.1 | 2 | 309 | |||
GenBank mRNA | BC166551.1 | - | 2 | 271 | 7307233 | |
FQ213977.1 | - | 2 | 237 | |||
FQ229926.1 | - | 2 | 225 |
