siRNA Details
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019250.2 | NP_062123.2 | 18 | 2772 | ||
XM_006233750.2 | XP_006233812.1 | 18 | 3317 | |||
XM_006233751.2 | XP_006233813.1 | 18 | 5250 | |||
XM_006233752.2 | XP_006233814.1 | 18 | 2830 | |||
XM_006233753.2 | XP_006233815.1 | 18 | 2794 | |||
XM_008761632.1 | XP_008759854.1 | 18 | 3095 | |||
GenBank mRNA | L07925.1 | - | 18 | 2772 |
