siRNA Details
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004230.1 | NP_001004230.1 | 7 | 889 | ||
GenBank mRNA | BC079029.1 | - | 7 | 889 | 7114041 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004230.1 | NP_001004230.1 | 7 | 889 | ||
GenBank mRNA | BC079029.1 | - | 7 | 889 | 7114041 |