siRNA Details
Gene Symbol: LOC100911434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762451.1 | XP_008760673.1 | 15 | 1879 |
Gene Symbol: Fer1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106534.2 | NP_001100004.2 | 16 | 1912 |
