siRNA Details
Gene Symbol: Ctsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011959.1 | NP_001011959.1 | 3, 4 | 515 | ||
XM_006235534.1 | XP_006235596.1 | 4, 5 | 773 | |||
GenBank mRNA | BC078934.1 | - | 3, 4 | 515 | 7108104 | |
FQ229081.1 | - | 3, 4 | 669 |
