siRNA Details
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106544.1 | NP_001100014.1 | 9 | 783 | ||
GenBank mRNA | FQ213909.1 | - | 9 | 761 | ||
FQ214139.1 | - | 9 | 766 | |||
FQ216824.1 | - | 9 | 766 | |||
FQ219919.1 | - | 9 | 766 |
