siRNA Details
Gene Symbol: RGD1565819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171096.1 | NP_001164567.1 | 1 | 2591 | ||
XM_006235714.2 | XP_006235776.1 | 2 | 3021 | |||
XM_008762455.1 | XP_008760677.1 | 2 | 3749 |
