siRNA Details
Gene Symbol: Helz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762606.1 | XP_008760828.1 | 9 | 6740 | ||
XM_008762607.1 | XP_008760829.1 | 9 | 6740 | |||
XM_230961.7 | XP_230961.5 | 9 | 6740 |
