siRNA Details
Gene Symbol: Hat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009657.1 | NP_001009657.1 | 7, 8 | 752 | ||
GenBank mRNA | BC087663.1 | - | 7, 8 | 752 | 7315259 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026115, AK169315, BC055460, AK010715, AK018270, and others.
Gene Ontology Categories:
