siRNA Details
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106567.1 | NP_001100037.1 | 6, 7 | 750 | ||
XM_006233783.2 | XP_006233845.1 | 7, 8 | 1916 |

More Information
Other Species Targeted

Gene Ontology Categories:
