siRNA Details
Gene Symbol: Prrc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234005.1 | XP_006234067.1 | 31 | 6657 | ||
XM_008761780.1 | XP_008760002.1 | 30 | 6725 | |||
XM_008761781.1 | XP_008760003.1 | 31 | 6788 | |||
XM_008761782.1 | XP_008760004.1 | 31 | 6784 | |||
XM_008761783.1 | XP_008760005.1 | 30 | 6722 | |||
XM_008761785.1 | XP_008760007.1 | 29 | 4655 |
