siRNA Details
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017278.1 | NP_058974.1 | 8, 9 | 716 | ||
GenBank mRNA | BC062233.1 | - | 8, 9 | 713 | 6919336 | |
D90265.1 | - | 8, 9 | 716 | |||
FQ212620.1 | - | 8, 9 | 685 | |||
FQ213606.1 | - | 8, 9 | 706 | |||
FQ215179.1 | - | 8, 9 | 695 | |||
FQ215378.1 | - | 8, 9 | 699 | |||
FQ216107.1 | - | 8, 9 | 709 | |||
FQ218243.1 | - | 748 | ||||
FQ220925.1 | - | 8, 9 | 681 | |||
FQ222787.1 | - | 8, 9 | 709 | |||
FQ223714.1 | - | 8, 9 | 685 | |||
FQ225726.1 | - | 8, 9 | 667 | |||
FQ229288.1 | - | 8, 9 | 743 | |||
M29859.1 | - | 8, 9 | 716 |
