siRNA Details
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235892.2 | XP_006235954.1 | 5 | 978 | ||
XM_006235894.2 | XP_006235956.1 | 5 | 965 | |||
XM_006235895.1 | XP_006235957.1 | 5 | 962 | |||
XM_006235896.2 | XP_006235958.1 | 5 | 979 | |||
GenBank mRNA | BC083715.1 | - | 5 | 982 | 7190690 | |
FQ215763.1 | - | 6 | 840 | |||
FQ228972.1 | - | 5 | 901 |
