siRNA Details
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106577.1 | NP_001100047.1 | 8 | 939 | ||
XM_008762640.1 | XP_008760862.1 | 8 | 894 | |||
XM_008762641.1 | XP_008760863.1 | 7 | 2477 | |||
XM_008762642.1 | XP_008760864.1 | 8 | 2489 | |||
XM_008762643.1 | XP_008760865.1 | 7 | 791 | |||
XM_008762644.1 | XP_008760866.1 | 8 | 2489 |
