siRNA Details
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019259.2 | NP_062132.2 | 2 | 293 | ||
GenBank mRNA | AJ001102.1 | - | 2 | 334 | ||
BC070510.1 | - | 2 | 293 | 7100505 | ||
FQ212622.1 | - | 2 | 318 | |||
FQ214537.1 | - | 2 | 343 | |||
FQ219796.1 | - | 2 | 316 | |||
FQ220621.1 | - | 2 | 336 |
