siRNA Details
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019259.2 | NP_062132.2 | 4 | 509 | ||
GenBank mRNA | BC070510.1 | - | 4 | 509 | 7100505 | |
FQ212622.1 | - | 4 | 534 | |||
FQ214537.1 | - | 4 | 559 | |||
FQ219796.1 | - | 4 | 532 | |||
FQ220621.1 | - | 4 | 552 |
