siRNA Details
Gene Symbol: Rbm48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024246.1 | NP_001019417.1 | 3 | 361 | ||
GenBank mRNA | BC093393.1 | - | 3 | 361 | 7388615 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024246.1 | NP_001019417.1 | 3 | 361 | ||
GenBank mRNA | BC093393.1 | - | 3 | 361 | 7388615 |