siRNA Details
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106580.2 | NP_001100050.1 | 8 | 1281 | ||
XM_006236029.2 | XP_006236091.1 | 8 | 1334 | |||
XM_006236030.2 | XP_006236092.1 | 7 | 1237 | |||
GenBank mRNA | BC168741.1 | - | 8 | 1281 | 6919194 |
