siRNA Details
Gene Symbol: Aass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100963.1 | NP_001094433.1 | 21 | 2373 | ||
GenBank mRNA | BC128771.1 | - | 21 | 2373 | 7130083 | |
FQ230063.1 | - | 2 | 84 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100963.1 | NP_001094433.1 | 21 | 2373 | ||
GenBank mRNA | BC128771.1 | - | 21 | 2373 | 7130083 | |
FQ230063.1 | - | 2 | 84 |