siRNA Details
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106590.1 | NP_001100060.1 | 3 | 493 | ||
XM_008762768.1 | XP_008760990.1 | 2 | 392 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106590.1 | NP_001100060.1 | 3 | 493 | ||
XM_008762768.1 | XP_008760990.1 | 2 | 392 |