siRNA Details
Gene Symbol: Kel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191611.1 | NP_001178540.1 | 10, 11 | 1267 | ||
XM_006236377.2 | XP_006236439.1 | 11, 12 | 1493 | |||
XM_006236378.1 | XP_006236440.1 | 11, 12 | 1493 | |||
XM_006236379.2 | XP_006236441.1 | 9, 10 | 1342 | |||
XM_006236380.2 | XP_006236442.1 | 11, 12 | 1493 | |||
XM_006236381.2 | XP_006236443.1 | 9, 10 | 1241 | |||
XM_006236382.1 | XP_006236444.1 | 9, 10 | 1214 | |||
XM_008762879.1 | XP_008761101.1 | 10, 11 | 1346 |
