siRNA Details
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009658.1 | NP_001009658.1 | 3 | 329 | ||
XM_006236625.2 | XP_006236687.1 | 459 | ||||
XM_006236626.2 | XP_006236688.1 | 459 | ||||
GenBank mRNA | BC088454.1 | - | 3 | 329 | 7127050 |
