siRNA Details
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106597.1 | NP_001100067.1 | 6 | 842 | ||
XM_006236659.2 | XP_006236721.1 | 7 | 1871 | |||
GenBank mRNA | BC166516.1 | - | 1300 | 7131845 |

More Information
Other Species Targeted

Gene Ontology Categories:
