siRNA Details
Gene Symbol: Kcnab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017303.2 | NP_058999.1 | 7 | 1142 | ||
XM_008761046.1 | XP_008759268.1 | 7 | 2413 | |||
GenBank mRNA | BC089219.1 | - | 7 | 1142 | 7320941 | |
X70662.1 | - | 7 | 816 |
