siRNA Details
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022926.1 | NP_075215.1 | 2 | 290 | ||
XM_006253083.1 | XP_006253145.1 | 2 | 315 | |||
GenBank mRNA | AF076167.1 | - | 2 | 290 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144731, NM_001167981, BC052461, AK041791, AK036523, and others.
Gene Ontology Categories:
