siRNA Details
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022926.1 | NP_075215.1 | 5, 6 | 1010 | ||
GenBank mRNA | AF076167.1 | - | 5, 6 | 1010 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144731, BC052461, AK151424, AK151753, BC049907, and others.
Gene Ontology Categories:
