siRNA Details
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106614.1 | NP_001100084.1 | 16 | 2686 | ||
XM_006237012.2 | XP_006237074.1 | 15 | 2353 | |||
XM_006237013.2 | XP_006237075.1 | 13 | 6472 | |||
XM_006237014.2 | XP_006237076.1 | 14 | 6454 | |||
XM_006237015.1 | XP_006237077.1 | 15 | 2115 | |||
XM_006237016.2 | XP_006237078.1 | 14 | 2314 | |||
XM_006237017.2 | XP_006237079.1 | 13 | 6415 | |||
XM_006237018.2 | XP_006237080.1 | 14 | 2257 | |||
XM_006237020.2 | XP_006237082.1 | 14 | 3377 | |||
XM_006237021.2 | XP_006237083.1 | 15 | 2298 | |||
XM_008763184.1 | XP_008761406.1 | 14 | 6511 | |||
XM_008763185.1 | XP_008761407.1 | 15 | 5712 | |||
XM_008763186.1 | XP_008761408.1 | 15 | 3203 | |||
XM_008763187.1 | XP_008761409.1 | 14 | 3241 | |||
XM_008763188.1 | XP_008761410.1 | 15 | 3147 | |||
XM_008763189.1 | XP_008761411.1 | 12 | 1907 | |||
GenBank mRNA | FQ223178.1 | - | 15 | 2143 |

More Information
Other Species Targeted

