siRNA Details
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106616.1 | NP_001100086.1 | 4, 5 | 1356 | ||
XM_006237181.2 | XP_006237243.1 | 4, 5 | 1129 | |||
XM_006237182.2 | XP_006237244.1 | 4, 5 | 1370 | |||
GenBank mRNA | FQ221326.1 | - | 4, 5 | 1268 |
