siRNA Details
Gene Symbol: Usp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106619.1 | NP_001100089.1 | 13, 14 | 1742 | ||
XM_006237338.1 | XP_006237400.1 | 13, 14 | 1735 |
Gene Symbol: LOC100911959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244623.2 | XP_006244685.1 | 7, 8 | 1085 | ||
XM_006244624.2 | XP_006244686.1 | 7, 8 | 1084 |
