siRNA Details
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106626.1 | NP_001100096.1 | 5, 6 | 698 | ||
XM_006237595.2 | XP_006237657.1 | 7, 8 | 1414 | |||
XM_008763376.1 | XP_008761598.1 | 7, 8 | 1394 | |||
XM_008763377.1 | XP_008761599.1 | 7, 8 | 1415 |
