siRNA Details
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106627.3 | NP_001100097.2 | 10, 11 | 1161 | ||
XM_008763378.1 | XP_008761600.1 | 6, 7 | 1598 | |||
GenBank mRNA | BC160907.1 | - | 10, 11 | 1161 | 7455489 | |
CB695629.1 | - | 2, 3 | 319 |
