siRNA Details
Gene Symbol: Arid1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106635.1 | NP_001100105.1 | 19 | 4937 | ||
XM_006239011.2 | XP_006239073.1 | 20 | 6134 | |||
XM_006239012.2 | XP_006239074.1 | 20 | 6131 |
