siRNA Details
Gene Symbol: Slc26a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106638.1 | NP_001100108.1 | 7 | 873 | ||
XM_006237911.2 | XP_006237973.1 | 8 | 1285 | |||
XM_006237912.2 | XP_006237974.1 | 8 | 1193 | |||
XM_006237913.2 | XP_006237975.1 | 7 | 1096 | |||
XM_008763530.1 | XP_008761752.1 | 8 | 1286 | |||
XM_008763531.1 | XP_008761753.1 | 7 | 1120 | |||
XM_008763532.1 | XP_008761754.1 | 8 | 1286 |
