siRNA Details
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024757.1 | NP_001019928.1 | 9 | 1197 | ||
XM_006237930.2 | XP_006237992.1 | 9 | 1112 | |||
XM_006237931.2 | XP_006237993.1 | 8 | 1539 | |||
XM_008763590.1 | XP_008761812.1 | 10 | 1410 | |||
XM_008763591.1 | XP_008761813.1 | 2 | 605 | |||
GenBank mRNA | BC097386.1 | - | 9 | 1197 | 7446512 |

More Information
Other Species Targeted

Gene Ontology Categories:
