siRNA Details
Gene Symbol: Fhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013088.1 | NP_001013106.1 | 6 | 832 | ||
XM_008763594.1 | XP_008761816.1 | 6 | 1197 | |||
XM_008763595.1 | XP_008761817.1 | 5 | 1135 | |||
GenBank mRNA | BC079327.1 | - | 6 | 832 | 7135490 |
